Anti-WNT10B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306582EA 499 EUR
ORIGTA306582
Anti-WNT10B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: WNT10B (wingless-type MMTV integration site family, member 10B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR