Anti-SLC35D1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306594EA 595 EUR
ORIGTA306594
Anti-SLC35D1 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1-2 μg/ml

Type: Primary
Antigen: SLC35D1 (solute carrier family 35 (UDP-GlcA/UDP-GalNAc transporter), member D1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR