Anti-METTL7A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306679EA 494 EUR
ORIGTA306679
Anti-METTL7A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 2 ?g/ml

Type: Primary
Antigen: METTL7A (methyltransferase like 7A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR