Anti-NIPSNAP3A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306692EA 549 EUR
ORIGTA306692
Anti-NIPSNAP3A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: NIPSNAP3A (nipsnap homolog 3A (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR