Anti-ZBTB1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306699EA 544 EUR
ORIGTA306699
Anti-ZBTB1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: ZBTB1 (zinc finger and BTB domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR