Anti-C12orf75 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306709EA 600 EUR
ORIGTA306709
Anti-C12orf75 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: C12orf75 (chromosome 12 open reading frame 75)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR