Anti-SPG11 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306723EA 499 EUR
ORIGTA306723
Anti-SPG11 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: SPG11 (spastic paraplegia 11 (autosomal recessive))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR