Anti-OTUD4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306742EA 600 EUR
ORIGTA306742
Anti-OTUD4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: OTUD4 (OTU deubiquitinase 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR