Anti-L3MBTL1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA308637EA 655 EUR
ORIGTA308637
Anti-L3MBTL1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: L3MBTL1 (l(3)mbt-like 1 (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR