Anti-BAAT Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA308660EA 440 EUR
ORIGTA308660
Anti-BAAT Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: BAAT (bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR