Anti-IDH2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA308664EA 544 EUR
ORIGTA308664
Anti-IDH2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: IDH2 (isocitrate dehydrogenase 2 (NADP+), mitochondrial)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR