Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NPR3 Rabbit Polyclonal Antibody

Supplier: OriGene
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:5000-1:20000

Type: Primary
Antigen: NPR3 (natriuretic peptide receptor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µl 100 µl ORIGTA308692
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol NPR3
Antigen name Natriuretic peptide receptor 3
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Amino acid number 265 to 514
Cross adsorption none
Immunogen Protein
Molecular weight 60 kDa
Purification purified by antigen affinity chromatography
Form Liquid
Storage buffer 0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty