Anti-FHL5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA308694EA 655 EUR
ORIGTA308694
Anti-FHL5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: FHL5 (four and a half LIM domains 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR