Anti-HRH3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA308704EA 655 EUR
ORIGTA308704
Anti-HRH3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot:1:1000-1:10000

Type: Primary
Antigen: HRH3 (histamine receptor H3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR