Anti-PPP2R3B Rabbit Polyclonal Antibody

ORIGTA315385EA 388 EUR
ORIGTA315385
Anti-PPP2R3B Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: PPP2R3B (protein phosphatase 2, regulatory subunit B'', beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR