Anti-TAS2R3 Rabbit Polyclonal Antibody

ORIGTA315425EA 388 EUR
ORIGTA315425
Anti-TAS2R3 Rabbit Polyclonal Antibody
  • Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: TAS2R3 (taste receptor, type 2, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR