Anti-TAS2R8 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315428EA 564 EUR
ORIGTA315428
Anti-TAS2R8 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: TAS2R8 (taste receptor, type 2, member 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR