Anti-LGR5 Mouse Antibody

Supplier: OriGene
ORIGTA400007EA 637 EUR
ORIGTA400007
Anti-LGR5 Mouse Antibody
Antibodies
Type: Primary
Antigen: LGR5 (leucine-rich repeat containing G protein-coupled receptor 5)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR