Anti-MYC Mouse Monoclonal Antibody [clone: OTI3F2]

Supplier: OriGene
ORIGTA500003EA 603 EUR
ORIGTA500003
Anti-MYC Mouse Monoclonal Antibody [clone: OTI3F2]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR