Anti-DFFA Mouse Monoclonal Antibody [clone: OTI5C4]

Supplier: OriGene
TA500062
ORIGTA500062EA 621 EUR
ORIGTA500062
Anti-DFFA Mouse Monoclonal Antibody [clone: OTI5C4]
Antibodies
Recommended Dilutions: Western Blot: 1:2500-5000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: DFFA (DNA fragmentation factor, 45kDa, alpha polypeptide)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR