Anti-TNNI3 Mouse Monoclonal Antibody [clone: OTI4A3]

Supplier: OriGene
ORIGTA500256EA 603 EUR
ORIGTA500256
Anti-TNNI3 Mouse Monoclonal Antibody [clone: OTI4A3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR