Anti-MAPK8 Mouse Monoclonal Antibody [clone: OTI2F10]

Supplier: OriGene
ORIGTA500275EA 608 EUR
ORIGTA500275
Anti-MAPK8 Mouse Monoclonal Antibody [clone: OTI2F10]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAPK8 (mitogen-activated protein kinase 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR