Anti-VEGFA Mouse Monoclonal Antibody [clone: OTI2F7]

Supplier: OriGene
ORIGTA500288EA 603 EUR
ORIGTA500288
Anti-VEGFA Mouse Monoclonal Antibody [clone: OTI2F7]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR