Anti-VEGFA Mouse Monoclonal Antibody [clone: OTI4E3]

Supplier: OriGene
ORIGTA500289EA 603 EUR
ORIGTA500289
Anti-VEGFA Mouse Monoclonal Antibody [clone: OTI4E3]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR