Anti-NEUROG1 Mouse Monoclonal Antibody [clone: OTI9A8]

Supplier: OriGene
ORIGTA500308EA 603 EUR
ORIGTA500308
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: OTI9A8]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: NEUROG1 (neurogenin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR