Anti-USP13 Mouse Monoclonal Antibody [clone: OTI4G11]

Supplier: OriGene
ORIGTA500318EA 608 EUR
ORIGTA500318
Anti-USP13 Mouse Monoclonal Antibody [clone: OTI4G11]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: USP13 (ubiquitin specific peptidase 13 (isopeptidase T-3))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR