Anti-GAD1 Mouse Monoclonal Antibody [clone: OTI5D8]

Supplier: OriGene
TA500330
ORIGTA500330EA 616 EUR
ORIGTA500330
Anti-GAD1 Mouse Monoclonal Antibody [clone: OTI5D8]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunoprecipitation: 4?g/ml

Type: Primary
Antigen: GAD1 (glutamate decarboxylase 1 (brain, 67kDa))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR