Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI7C5]

Supplier: OriGene
TA500442
ORIGTA500442EA 616 EUR
ORIGTA500442
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI7C5]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1 (nuclear receptor binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR