Anti-CD4 Mouse Monoclonal Antibody [clone: OTI4A11]

Supplier: OriGene
ORIGTA500479EA 581 EUR
ORIGTA500479
Anti-CD4 Mouse Monoclonal Antibody [clone: OTI4A11]
Antibodies
  • Recommended Dilutions: Western Blot: 1:200-1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR