Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI7E7]

Supplier: OriGene
ORIGTA500486EA 603 EUR
ORIGTA500486
Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI7E7]
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:50-100; Immunoprecipitation: 2μg/500μl

Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR