Anti-MRI1 Mouse Monoclonal Antibody [clone: OTI6A10]

Supplier: OriGene
ORIGTA500490EA 603 EUR
ORIGTA500490
Anti-MRI1 Mouse Monoclonal Antibody [clone: OTI6A10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100

Type: Primary
Antigen: MRI1 (methylthioribose-1-phosphate isomerase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR