Anti-DHFR Mouse Monoclonal Antibody [clone: OTI3A11]

Supplier: OriGene
ORIGTA500517EA 603 EUR
ORIGTA500517
Anti-DHFR Mouse Monoclonal Antibody [clone: OTI3A11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: DHFR (dihydrofolate reductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR