Anti-BUB1B Mouse Monoclonal Antibody [clone: OTI5D9]

Supplier: OriGene
TA500533
ORIGTA500533EA 616 EUR
ORIGTA500533
Anti-BUB1B Mouse Monoclonal Antibody [clone: OTI5D9]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Immunofluorescence: 1:100

Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR