Anti-NTRK3 Mouse Monoclonal Antibody [clone: OTI24B3]

Supplier: OriGene
ORIGTA500536EA 603 EUR
ORIGTA500536
Anti-NTRK3 Mouse Monoclonal Antibody [clone: OTI24B3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: NTRK3 (neurotrophic tyrosine kinase, receptor, type 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR