Anti-FRK Mouse Monoclonal Antibody [clone: OTI7A7]

Supplier: OriGene
ORIGTA500549EA 603 EUR
ORIGTA500549
Anti-FRK Mouse Monoclonal Antibody [clone: OTI7A7]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: FRK (fyn-related kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR