Anti-SSX2 Mouse Monoclonal Antibody [clone: OTI4D10]

Supplier: OriGene
ORIGTA500620EA 603 EUR
ORIGTA500620
Anti-SSX2 Mouse Monoclonal Antibody [clone: OTI4D10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: SSX2 (synovial sarcoma, X breakpoint 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR