Anti-PDE10A Mouse Monoclonal Antibody [clone: OTI1B2]

Supplier: OriGene
ORIGTA500635EA 608 EUR
ORIGTA500635
Anti-PDE10A Mouse Monoclonal Antibody [clone: OTI1B2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: PDE10A (phosphodiesterase 10A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR