Anti-GBP2 Mouse Monoclonal Antibody [clone: OTI5C8]

Supplier: OriGene
TA500640
ORIGTA500640EA 593 EUR
ORIGTA500640
Anti-GBP2 Mouse Monoclonal Antibody [clone: OTI5C8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP2 (guanylate binding protein 2, interferon-inducible)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR