Anti-HSD17B10 Mouse Monoclonal Antibody [clone: OTI3B5]

Supplier: OriGene
ORIGTA500687EA 603 EUR
ORIGTA500687
Anti-HSD17B10 Mouse Monoclonal Antibody [clone: OTI3B5]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HSD17B10 (hydroxysteroid (17-beta) dehydrogenase 10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR