Anti-PRL Mouse Monoclonal Antibody [clone: OTI2B7]

Supplier: OriGene
ORIGTA500721EA 581 EUR
ORIGTA500721
Anti-PRL Mouse Monoclonal Antibody [clone: OTI2B7]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PRL (prolactin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR