Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI6B11]

Supplier: OriGene
ORIGTA500722EA 603 EUR
ORIGTA500722
Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI6B11]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B (ring finger protein 144B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR