Anti-CYP2E1 Mouse Monoclonal Antibody [clone: OTI5B9]

Supplier: OriGene
ORIGTA500734EA 603 EUR
ORIGTA500734
Anti-CYP2E1 Mouse Monoclonal Antibody [clone: OTI5B9]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CYP2E1 (cytochrome P450, family 2, subfamily E, polypeptide 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR