Anti-IDH3A Mouse Monoclonal Antibody [clone: OTI2F11]

Supplier: OriGene
ORIGTA500739EA 621 EUR
ORIGTA500739
Anti-IDH3A Mouse Monoclonal Antibody [clone: OTI2F11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: IDH3A (isocitrate dehydrogenase 3 (NAD+) alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR