Anti-AKR1A1 Mouse Monoclonal Antibody [clone: OTI10E11]

Supplier: OriGene
ORIGTA500744EA 608 EUR
ORIGTA500744
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: OTI10E11]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: AKR1A1 (aldo-keto reductase family 1, member A1 (aldehyde reductase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR