Anti-HDAC10 Mouse Monoclonal Antibody [clone: OTI8G5]

Supplier: OriGene
ORIGTA500751EA 581 EUR
ORIGTA500751
Anti-HDAC10 Mouse Monoclonal Antibody [clone: OTI8G5]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: HDAC10 (histone deacetylase 10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR