Anti-BIRC7 Mouse Monoclonal Antibody [clone: OTI1B10]

Supplier: OriGene
ORIGTA500761EA 608 EUR
ORIGTA500761
Anti-BIRC7 Mouse Monoclonal Antibody [clone: OTI1B10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: BIRC7 (baculoviral IAP repeat containing 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR