Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI8D9]

Supplier: OriGene
ORIGTA500783EA 603 EUR
ORIGTA500783
Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI8D9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR