Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI1D3]

Supplier: OriGene
ORIGTA500798EA 581 EUR
ORIGTA500798
Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI1D3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TTLL12 (tubulin tyrosine ligase-like family, member 12)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR