Anti-CLDN5 Mouse Monoclonal Antibody [clone: OTI1G4]

Supplier: OriGene
ORIGTA500843EA 608 EUR
ORIGTA500843
Anti-CLDN5 Mouse Monoclonal Antibody [clone: OTI1G4]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CLDN5 (claudin 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR