Anti-GRIPAP1 Mouse Monoclonal Antibody [clone: OTI1F4]

Supplier: OriGene
ORIGTA500854EA 581 EUR
ORIGTA500854
Anti-GRIPAP1 Mouse Monoclonal Antibody [clone: OTI1F4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GRIPAP1 (GRIP1 associated protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR